missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Amino-2-naphthoic acid, 85%, tech., ACROS Organics™

Manufacturer:  Acros Organics 104140250

 View more versions of this product

Catalog No. AC104140250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3-Amino-2-naphthoic acid
Glass bottle
210.0°C
85%
80.0
85%
H2NC10H6CO2H
14, 535
3-amino-2-naphthoic acid, 2-amino-3-naphthoic acid, 3-aminoisonaphthoic acid, 2-naphthalenecarboxylic acid, 3-amino, 2-amino-3-carboxynaphthalene, 2-naphthoic acid, 3-amino, 3-amino-2-naphthalenecarboxylic acid, unii-iv5z0phl5d, iv5z0phl5d, 3-amino-naphthalene-2-carboxylic acid
C1=CC=C2C=C(C(=CC2=C1)C(=O)O)N
187.198
187.2
85%
Authentic
Green to Green-Yellow
25g
5959-52-4
100.0
C11H9NO2
MFCD00004115
15, 448
XFXOLBNQYFRSLQ-UHFFFAOYSA-N
3-aminonaphthalene-2-carboxylic acid
22244
Powder
Technical

Chemical Identifiers

5959-52-4
187.198
XFXOLBNQYFRSLQ-UHFFFAOYSA-N
22244
C1=CC=C2C=C(C(=CC2=C1)C(=O)O)N
C11H9NO2
MFCD00004115
3-amino-2-naphthoic acid, 2-amino-3-naphthoic acid, 3-aminoisonaphthoic acid, 2-naphthalenecarboxylic acid, 3-amino, 2-amino-3-carboxynaphthalene, 2-naphthoic acid, 3-amino, 3-amino-2-naphthalenecarboxylic acid, unii-iv5z0phl5d, iv5z0phl5d, 3-amino-naphthalene-2-carboxylic acid
3-aminonaphthalene-2-carboxylic acid
Safety and Handling

Safety and Handling

GHS P Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 227-726-2

RTECSNumber : QL1400000

TSCA : TSCA

SDS Documents