Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Amino-2-methylphenol, 98%

$20.64 - $20.64

Chemical Identifiers

Chemical Name or Material 3-Amino-2-methylphenol
CAS 53222-92-7
Formula Weight 123.16
MDL Number MFCD00007787
Beilstein 2716070
Synonym 3-Amino-o-cresol; 3-Hydroxy-2-methylaniline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6655403

 
alfa aesar™
H6655403
1g Each for $20.64

AAH6655406

 
alfa aesar™
H6655406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6655414

 
alfa aesar™
H6655414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 129° to 130°C
Formula Weight 123.16
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 3-Amino-2-methylphenol
Physical Form Solid
Assay 98%
Quantity 1g
MDL Number MFCD00007787
CAS 53222-92-7
Beilstein 2716070
Synonym 3-Amino-o-cresol; 3-Hydroxy-2-methylaniline
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.