missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Amino-2-methylbenzyl alcohol, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC440020250
Specifications
3-Amino-2-methylbenzyl alcohol | |
Glass bottle | |
108.9°C to 109.7°C | |
83647-42-1 | |
100.0 | |
C8H11NO | |
UVYZMJMDIMWDNJ-UHFFFAOYSA-N | |
(3-amino-2-methylphenyl)methanol | |
577175 | |
Powder |
Authentic | |
Orange-Purple to White | |
25g | |
96.0 | |
96% min. (GC) | |
3-amino-2-methyl benzyl alcohol, 3-amino-2-methyl-phenyl methanol, 3-amino-2-methylbenzenemethanol, 3-amino-2-methylbenzyl alcohol, 3-amino-2-methylbenzylalcohol, 3-amino-2-methylphenyl metanol, 3-amino-2-methylphenyl methanol, acmc-20a9oa, benzenemethanol, 3-amino-2-methyl, pubchem19790 | |
CC1=C(C=CC=C1N)CO | |
137.18 | |
137.18 | |
97% |
Chemical Identifiers
83647-42-1 | |
137.18 | |
3-amino-2-methyl benzyl alcohol, 3-amino-2-methyl-phenyl methanol, 3-amino-2-methylbenzenemethanol, 3-amino-2-methylbenzyl alcohol, 3-amino-2-methylbenzylalcohol, 3-amino-2-methylphenyl metanol, 3-amino-2-methylphenyl methanol, acmc-20a9oa, benzenemethanol, 3-amino-2-methyl, pubchem19790 | |
(3-amino-2-methylphenyl)methanol |
C8H11NO | |
UVYZMJMDIMWDNJ-UHFFFAOYSA-N | |
577175 | |
CC1=C(C=CC=C1N)CO |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e
GHS Signal Word: Warning