missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Amino-2-methylbenzyl alcohol, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC440020050

 View more versions of this product

Catalog No. AC440020050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3-Amino-2-methylbenzyl alcohol
Orange-Purple to White
5g
96.0
96% min. (GC)
3-amino-2-methyl benzyl alcohol, 3-amino-2-methyl-phenyl methanol, 3-amino-2-methylbenzenemethanol, 3-amino-2-methylbenzyl alcohol, 3-amino-2-methylbenzylalcohol, 3-amino-2-methylphenyl metanol, 3-amino-2-methylphenyl methanol, acmc-20a9oa, benzenemethanol, 3-amino-2-methyl, pubchem19790
CC1=C(C=CC=C1N)CO
137.18
137.18
97%
Authentic
108.9°C to 109.7°C
83647-42-1
100.0
C8H11NO
UVYZMJMDIMWDNJ-UHFFFAOYSA-N
(3-amino-2-methylphenyl)methanol
577175
Powder

Chemical Identifiers

83647-42-1
137.18
3-amino-2-methyl benzyl alcohol, 3-amino-2-methyl-phenyl methanol, 3-amino-2-methylbenzenemethanol, 3-amino-2-methylbenzyl alcohol, 3-amino-2-methylbenzylalcohol, 3-amino-2-methylphenyl metanol, 3-amino-2-methylphenyl methanol, acmc-20a9oa, benzenemethanol, 3-amino-2-methyl, pubchem19790
(3-amino-2-methylphenyl)methanol
C8H11NO
UVYZMJMDIMWDNJ-UHFFFAOYSA-N
577175
CC1=C(C=CC=C1N)CO
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e

GHS Signal Word: Warning

Documents