missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Amino-2-methoxypyridine-5-boronic acid pinacol ester, 96%

Manufacturer:  Alfa Aesar™ H5457003

 View more versions of this product

Catalog No. AAH5457003



Description

Description

Applications
It is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Incompatible with oxidizing agents, air, heat. Refrigerated (2-4°C).
Specifications

Specifications

3-Amino-2-methoxypyridine-5-boronic acid pinacol ester
1g
C12H19BN2O3
2-methoxy-3-aminopyridine-5-boronic acid pinacol ester
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(N=C2)OC)N
250.105
250.1
131°C to 133°C
893440-50-1
MFCD12923427
KYYKGOURQXPERA-UHFFFAOYSA-N
2-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-3-amine
59557920
96%

Chemical Identifiers

893440-50-1
250.105
KYYKGOURQXPERA-UHFFFAOYSA-N
59557920
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(N=C2)OC)N
C12H19BN2O3
MFCD12923427
2-methoxy-3-aminopyridine-5-boronic acid pinacol ester
2-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-3-amine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents