missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Amino-2-methoxypyridine-4-carboxylic acid, 95%

$171.18 - $507.35

Chemical Identifiers

CAS 870997-81-2
Molecular Formula C7H8N2O3
Molecular Weight (g/mol) 168.152
MDL Number MFCD18262266
InChI Key ABVDQGNEDSKBLA-UHFFFAOYSA-N
Synonym 3-amino-2-methoxyisonicotinic acid, 3-amino-2-methoxy-4-pyridinecarboxylic acid, 3-amino-2-methoxyisonicotinicacid, 3-amino-2-methoxy-isonicotinic acid
PubChem CID 53634618
IUPAC Name 3-amino-2-methoxypyridine-4-carboxylic acid
SMILES COC1=NC=CC(=C1N)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH66289MD View Documents Alfa Aesar™
H66289MD
250mg Each for $171.18
 
AAH6628903 View Documents Alfa Aesar™
H6628903
1g Each for $507.35
 
Specifications

Chemical Identifiers

870997-81-2
168.152
ABVDQGNEDSKBLA-UHFFFAOYSA-N
53634618
COC1=NC=CC(=C1N)C(=O)O
C7H8N2O3
MFCD18262266
3-amino-2-methoxyisonicotinic acid, 3-amino-2-methoxy-4-pyridinecarboxylic acid, 3-amino-2-methoxyisonicotinicacid, 3-amino-2-methoxy-isonicotinic acid
3-amino-2-methoxypyridine-4-carboxylic acid

Specifications

250mg
C7H8N2O3
19162520
ABVDQGNEDSKBLA-UHFFFAOYSA-N
3-amino-2-methoxypyridine-4-carboxylic acid
53634618
Solid
3-Amino-2-methoxypyridine-4-carboxylic acid
870997-81-2
MFCD18262266
3-amino-2-methoxyisonicotinic acid, 3-amino-2-methoxy-4-pyridinecarboxylic acid, 3-amino-2-methoxyisonicotinicacid, 3-amino-2-methoxy-isonicotinic acid
COC1=NC=CC(=C1N)C(=O)O
168.152
168.15
95%