Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Amino-2-methoxy-6-methylpyridine, 97%

Chemical Identifiers

Chemical Name or Material 3-Amino-2-methoxy-6-methylpyridine
CAS 186413-79-6
Formula Weight 138.17g/mol
MDL Number MFCD03095079
Synonym 2-Methoxy-6-methylpyridin-3-amine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6131003

 
alfa aesar™
H61310-03
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6131006

 
alfa aesar™
H61310-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Color Dark brown
Quantity 1g
Chemical Name or Material 3-Amino-2-methoxy-6-methylpyridine
CAS 186413-79-6
Physical Form Liquid
Assay 97%
Formula Weight 138.17g/mol
MDL Number MFCD03095079
Synonym 2-Methoxy-6-methylpyridin-3-amine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.