Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Amino-2-fluoro-4-methylpyridine, 98%

Chemical Identifiers

Chemical Name or Material 3-Amino-2-fluoro-4-methylpyridine
CAS 173435-32-0
Formula Weight 126.13
MDL Number MFCD04112556
Beilstein 13755262
Synonym 3-Amino-2-fluoro-4-picoline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6475803

  SDS
alfa aesar™
H6475803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6475806

  SDS
alfa aesar™
H6475806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH64758MD

  SDS
alfa aesar™
H64758MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 126.13
Melting Point 63° to 64°C
Health Hazard 2 GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Beilstein 13755262
Assay 98%
Chemical Name or Material 3-Amino-2-fluoro-4-methylpyridine
Health Hazard 1 Warning
Color Off-white
Synonym 3-Amino-2-fluoro-4-picoline
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Physical Form Solid
MDL Number MFCD04112556
CAS 173435-32-0
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.