missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Amino-2-chloro-5-(trifluoromethyl)pyridine, 98%

Manufacturer:  Alfa Aesar™ H6494506

 View more versions of this product

Catalog No. AAH6494506



Description

Description

Applications
It’s employed as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Specifications

Specifications

3-Amino-2-chloro-5-(trifluoromethyl)pyridine
5g
C6H4ClF3N2
19757170
Slightly soluble in water.
C1=C(C=NC(=C1N)Cl)C(F)(F)F
196.557
196.56
98%
White to Brown
72587-18-9
MFCD07375383
2-chloro-3-amino-5-trifluoromethyl pyridine, 2-chloro-5-trifluoromethyl pyridin-3-amine, 2-chloro-5-trifluoromethyl-3-pyridinamine, 3-amino-2-chloro-5-trifluoromethyl pyridine, 3-amino-2-chloro-5-trifluoromethylpyridine
HABKYPVMMHBOIW-UHFFFAOYSA-N
2-chloro-5-(trifluoromethyl)pyridin-3-amine
12919757
Solid

Chemical Identifiers

72587-18-9
196.557
HABKYPVMMHBOIW-UHFFFAOYSA-N
12919757
C1=C(C=NC(=C1N)Cl)C(F)(F)F
C6H4ClF3N2
MFCD07375383
2-chloro-3-amino-5-trifluoromethyl pyridine, 2-chloro-5-trifluoromethyl pyridin-3-amine, 2-chloro-5-trifluoromethyl-3-pyridinamine, 3-amino-2-chloro-5-trifluoromethyl pyridine, 3-amino-2-chloro-5-trifluoromethylpyridine
2-chloro-5-(trifluoromethyl)pyridin-3-amine
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

Documents