Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Amino-2-chloro-4-methylpyridine, 97%

$68.35 - $261.68

Chemical Identifiers

Chemical Name or Material 3-Amino-2-chloro-4-methylpyridine
CAS 133627-45-9
Molecular Formula C6H7ClN2
Formula Weight 142.59
MDL Number MFCD00673152
Synonym 3-Amino-2-chloro-4-picoline; 2-Chloro-4-methyl-3-pyridinamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2605803

 
alfa aesar™
H2605803
1g Each for $68.35

AAH2605806

 
alfa aesar™
H2605806
5g Each for $261.68
Description & Specifications

Specifications

Formula Weight 142.59
Chemical Name or Material 3-Amino-2-chloro-4-methylpyridine
Melting Point 64° to 68°C
Quantity 1g
Assay 97%
Molecular Formula C6H7ClN2
MDL Number MFCD00673152
CAS 133627-45-9
Synonym 3-Amino-2-chloro-4-picoline; 2-Chloro-4-methyl-3-pyridinamine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.