Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Amino-2,6-dimethylpyridine, 97%


Quantity 5g

Manufacturer: alfa aesar™  L1984206

 View more versions of this product

Catalog No. AAL1984206

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 122.17
CAS 3430-33-9
Beilstein 2424
UN Number UN2811
MDL Number MFCD01646409
Molecular Formula C7H10N2
Chemical Name or Material 3-Amino-2,6-dimethylpyridine
Health Hazard 2 GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 97%
Health Hazard 1 Danger
Quantity 5g
Synonym 3-Amino-2,6-lutidine
Boiling Point 230°C
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Melting Point 121° to 123°C
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Specifications

Chemical Name or Material 3-Amino-2,6-dimethylpyridine
CAS 3430-33-9
Molecular Formula C7H10N2
Formula Weight 122.17
MDL Number MFCD01646409
UN Number UN2811
Beilstein 2424
Synonym 3-Amino-2,6-lutidine

Applications
3-Amino-2,6-dimethylpyridine is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Accessories

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents