Please login to your online account to display your discounted pricing

3-Amino-2,6-dimethoxypyridine monohydrochloride, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Amino-2, 6-dimethoxypyridine monohydrochloride
Name Note 95%
CAS 80789-72-6
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C7 H10 N2 O2 . H Cl
Linear Formula (CH3O)2(C5H2N)NH2
Formula Weight 190.63
MDL Number MFCD00012816
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC162320025

  SDS
fisher scientific acros
162320025
Glass bottle 2.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 190.63
Quantity 2.5g
TSCA TSCA
EINECS Number 279-554-2
CAS Min % 96.0
CAS Max % 100.0
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 80789-72-6
Name Note 95%
Linear Formula (CH3O)2(C5H2N)NH2
MDL Number MFCD00012816
Molecular Formula C7 H10 N2 O2 . H Cl
Assay Percent Range 97%
Chemical Name or Material 3-Amino-2, 6-dimethoxypyridine monohydrochloride
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.