Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Amino-2,6-difluorobenzoic acid, 95%

Chemical Identifiers

Chemical Name or Material 3-Amino-2,6-difluorobenzoic acid
CAS 83141-11-1
Formula Weight 173.12
MDL Number MFCD07368833
Beilstein 13330504
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6664503

 
alfa aesar™
H6664503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6664506

 
alfa aesar™
H6664506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 173.12
Physical Form Solid
MDL Number MFCD07368833
CAS 83141-11-1
Chemical Name or Material 3-Amino-2,6-difluorobenzoic acid
Assay 95%
Quantity 1g
Beilstein 13330504