Learn More
3-Amino-2,2,5,5-tetramethyl-1-pyrrolidinyloxy, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 401100020
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
3-Amino-2,2,5,5-tetramethyl-1-pyrrolidinyloxy | |
34272-83-8 | |
100.0 | |
Authentic | |
Glass bottle | |
MFCD00046088 | |
3s-1-hydroxy-2,2,5,5-tetramethylpyrrolidin-3-aminium | |
CC1(CC(C(N1O)(C)C)N)C | |
157.24 | |
157.24 |
99% | |
98.5 | |
75.0°C to 80.0°C (1.0mmHg) | |
99% | |
C8H17N2O | |
2g | |
USGBLEMQARVRDC-UHFFFAOYSA-N | |
1-hydroxy-2,2,5,5-tetramethylpyrrolidin-3-amine | |
558590 | |
99% |
Chemical Identifiers
34272-83-8 | |
157.24 | |
USGBLEMQARVRDC-UHFFFAOYSA-N | |
558590 | |
CC1(CC(C(N1O)(C)C)N)C |
C8H17N2O | |
MFCD00046088 | |
3s-1-hydroxy-2,2,5,5-tetramethylpyrrolidin-3-aminium | |
1-hydroxy-2,2,5,5-tetramethylpyrrolidin-3-amine |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breath
GHS Signal Word: Danger
TSCA : TSCA
RUO â Research Use Only