Please login to your online account to display your discounted pricing

3-Amino-2,2,5,5-tetramethyl-1-pyrrolidinyloxy 99%, ACROS Organics™


Quantity 500mg
Packaging Glass bottle

Manufacturer: fisher scientific acros  401105000

 View more versions of this product

Catalog No. AC401105000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Hygroscopic Crystals
Formula Weight 157.24
Quantity 500mg
Color Orange
TSCA TSCA
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 99%
CAS Min % 98.5
Name Note 99%
Molecular Formula C8H17N2O
Chemical Name or Material 3-Amino-2,2,5,5-tetramethyl-1-pyrrolidinyloxy
CAS 34272-83-8
MDL Number MFCD00046088
Packaging Glass bottle
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Causes serious eye damage.
Harmful if inhaled.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 3-Amino-2,2,5,5-tetramethyl-1-pyrrolidinyloxy
Name Note 99%
CAS 34272-83-8
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H17N2O
Formula Weight 157.24
MDL Number MFCD00046088
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Causes serious eye damage.
Harmful if inhaled.
Harmful if swallowed.
Causes severe skin burns and eye damage.

SDS & Safety Documents