missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Amino-5-hydroxypyrazole, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 271720250

 View more versions of this product

Catalog No. AC271720250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3-Amino-5-hydroxypyrazole
Glass bottle
25g
97.5
98%
MFCD00022384
1h-pyrazol-5-ol, 3-amino, 3-amino-1,2-dihydropyrazol-5-one, 3-amino-1h-pyrazol-5-ol, 3-amino-5-hydroxypyrazole, 3-pyrazolin-5-one, 3-amino, 5-amino-1h-pyrazol-3 2h-one, 5-amino-1h-pyrazol-3-ol, 5-amino-2,3-dihydro-1h-pyrazol-3-one, 5-amino-2h-pyrazol-3-ol, 5-aminopyrazol-3-ol
C1=C(NNC1=O)N
99.093
99.09
≥97.5%
Authentic
Beige-Brown to Beige-Yellow
6126-22-3
100.0
C3H5N3O
25, 3516
QZBGOTVBHYKUDS-UHFFFAOYSA-N
5-amino-1,2-dihydropyrazol-3-one
96221
Crystalline Powder

Chemical Identifiers

6126-22-3
99.093
QZBGOTVBHYKUDS-UHFFFAOYSA-N
96221
C1=C(NNC1=O)N
C3H5N3O
MFCD00022384
1h-pyrazol-5-ol, 3-amino, 3-amino-1,2-dihydropyrazol-5-one, 3-amino-1h-pyrazol-5-ol, 3-amino-5-hydroxypyrazole, 3-pyrazolin-5-one, 3-amino, 5-amino-1h-pyrazol-3 2h-one, 5-amino-1h-pyrazol-3-ol, 5-amino-2,3-dihydro-1h-pyrazol-3-one, 5-amino-2h-pyrazol-3-ol, 5-aminopyrazol-3-ol
5-amino-1,2-dihydropyrazol-3-one
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents