missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Amino-1H-indazole, 95%, Thermo Scientific™

Chemical Identifiers

CAS 874-05-5
Molecular Formula C7H7N3
Molecular Weight (g/mol) 133.15
MDL Number MFCD00182045
InChI Key YDTDKKULPWTHRV-UHFFFAOYSA-N
Synonym 1h-indazol-3-ylamine, 1h-indazole, 3-amino, 1h-indazole-3-ylamine, 2h-indazol-3-amine, 3-amino-1h-indazole, 3-aminoindazole, 3-indazolamine, amino-indazole, aminoindazole, indazole amine
PubChem CID 13399
IUPAC Name 1H-indazol-3-amine
SMILES C1=CC=C2C(=C1)C(=NN2)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC435740010
SDS View Documents Promotion Details
Thermo Fisher Scientific
435740010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC435740050
SDS View Documents Promotion Details
Thermo Fisher Scientific
435740050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

874-05-5
133.15
YDTDKKULPWTHRV-UHFFFAOYSA-N
13399
C1=CC=C2C(=C1)C(=NN2)N
C7H7N3
MFCD00182045
1h-indazol-3-ylamine, 1h-indazole, 3-amino, 1h-indazole-3-ylamine, 2h-indazol-3-amine, 3-amino-1h-indazole, 3-aminoindazole, 3-indazolamine, amino-indazole, aminoindazole, indazole amine
1H-indazol-3-amine

Specifications

874-05-5
100.0
156°C to 158°C
C7H7N3
MFCD00182045
1g
YDTDKKULPWTHRV-UHFFFAOYSA-N
1H-indazol-3-amine
13399
Liquid or Solid
3-Amino-1H-indazole
94.0
Colorless or White to Yellow
94% min. (HPLC)
Authentic
Glass bottle
1h-indazol-3-ylamine, 1h-indazole, 3-amino, 1h-indazole-3-ylamine, 2h-indazol-3-amine, 3-amino-1h-indazole, 3-aminoindazole, 3-indazolamine, amino-indazole, aminoindazole, indazole amine
C1=CC=C2C(=C1)C(=NN2)N
133.15
133.15
95%
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash

GHS Signal Word: Warning

SDS
Product Certifications

RUO – Research Use Only