missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Amino-1H-1,2,4-triazole, 96%, Alfa Aesar

Manufacturer:  Alfa Aesar A148710B

 View more versions of this product

Catalog No. AAA148710B



Description

Description

Applications
3-Amino-1,2,4-triazole is an inhibitor of mitochondrial and chloroplast function. Commercial grade 3- amino-1,2,4-triazole (which generally contains catalase anti-inhibitory impurities) is used as a cotton defoliant.

Solubility
It is soluble in water, methanol, ethanol and chloroform, and sparingly soluble in ethyl acetate. It is insoluble in ether and acetone.

Notes
Store desiccated at -20°C. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away from Strong oxidizing agents, Strong acids, Copper, Iron and iron salts.
Specifications

Specifications

3-Amino-1H-1,2,4-triazole
147°C to 153°C
61-82-5
C2H4N4
UN3077
14,489
KLSJWNVTNUYHDU-UHFFFAOYSA-N
1H-1,2,4-triazol-5-amine
1639
84.08
Odorless
1000g
96%
MFCD00005230
107687
3-amino-1,2,4-triazole, amitrole, aminotriazole, 4h-1,2,4-triazol-3-amine, 1h-1,2,4-triazol-3-amine, amitrol, amerol, amizol, emisol, vorox
C1=NNC(=N1)N
84.082
CHEBI:40036
96%

Chemical Identifiers

61-82-5
84.082
KLSJWNVTNUYHDU-UHFFFAOYSA-N
1639
1H-1,2,4-triazol-5-amine
C2H4N4
MFCD00005230
3-amino-1,2,4-triazole, amitrole, aminotriazole, 4h-1,2,4-triazol-3-amine, 1h-1,2,4-triazol-3-amine, amitrol, amerol, amizol, emisol, vorox
CHEBI:40036
C1=NNC(=N1)N
Safety and Handling

Safety and Handling

GHS H Statement
H361-H373
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
P260-P281-P308+P313-P314-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Use personal protective equipment as required.
IF exposed or concerned: Get medical advice/attention.
Get medical advice/attention if you feel unwell.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

RTECSNumber : XZ3850000

TSCA : TSCA

SDS Documents