Learn More
3-Amino-1H-1,2,4-triazole, 95%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 264571000
Specifications
3-Amino-1H-1, 2, 4-triazole | |
Glass bottle | |
149°C to 156°C | |
95% | |
94.0 | |
95% | |
MFCD00005230 | |
15, 485 | |
Solubility in water: 280g/L (20°C). Other solubilities: 520g/L (50°C),sparingly soluble in chloroform,dichloromethane,and acetonitrile,slightly soluble in ethylacetate,pract.insol. in diethylether,hydrocarbons,acetone | |
C1=NNC(=N1)N | |
84.08 | |
CHEBI:40036 | |
Powder or Flakes |
Authentic | |
White or Yellow | |
100g | |
61-82-5 | |
100.0 | |
C2H4N4 | |
26, 137 | |
1h-1,2,4-triazol-3-amine, 3-amino-1,2,4-triazole, 4h-1,2,4-triazol-3-amine, amerol, aminotriazole, amitrol, amitrole, amizol, emisol, vorox | |
KLSJWNVTNUYHDU-UHFFFAOYSA-N | |
1H-1,2,4-triazol-5-amine | |
1639 | |
84.08 | |
95% |
Chemical Identifiers
61-82-5 | |
84.08 | |
KLSJWNVTNUYHDU-UHFFFAOYSA-N | |
1639 | |
1H-1,2,4-triazol-5-amine |
C2H4N4 | |
MFCD00005230 | |
1h-1,2,4-triazol-3-amine, 3-amino-1,2,4-triazole, 4h-1,2,4-triazol-3-amine, amerol, aminotriazole, amitrol, amitrole, amizol, emisol, vorox | |
CHEBI:40036 | |
C1=NNC(=N1)N |
Safety and Handling
GHS H Statement:
May cause damage to organs through prolonged or repeated exposure.
Toxic to aquatic life with long lasting effects.
Suspected of damaging the unborn child.
GHS P Statement:
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 200-521-5
RTECSNumber : XZ3850000
TSCA : TSCA