missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Amino-1H-1,2,4-triazole, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC264570100

 View more versions of this product

Catalog No. AC264570100



Description

Description

Specifications

Specifications

3-Amino-1H-1, 2, 4-triazole
Glass bottle
149°C to 156°C
95%
94.0
95%
MFCD00005230,MFCD00053362
15, 485
Solubility in water: 280g/L (20°C). Other solubilities: 520g/L (50°C),sparingly soluble in chloroform,dichloromethane,and acetonitrile,slightly soluble in ethylacetate,pract.insol. in diethylether,hydrocarbons,acetone
NC1=NC=NN1
84.08
CHEBI:40036
Powder or Flakes
Authentic
White or Yellow
10g
61-82-5
100.0
C2H4N4
26, 137
1h-1,2,4-triazol-3-amine, 3-amino-1,2,4-triazole, 4h-1,2,4-triazol-3-amine, amerol, aminotriazole, amitrol, amitrole, amizol, emisol, vorox
KLSJWNVTNUYHDU-UHFFFAOYSA-N
1H-1,2,4-triazol-5-amine
1639
84.08
95%

Chemical Identifiers

61-82-5
84.08
KLSJWNVTNUYHDU-UHFFFAOYSA-N
1639
1H-1,2,4-triazol-5-amine
C2H4N4
MFCD00005230,MFCD00053362
1h-1,2,4-triazol-3-amine, 3-amino-1,2,4-triazole, 4h-1,2,4-triazol-3-amine, amerol, aminotriazole, amitrol, amitrole, amizol, emisol, vorox
CHEBI:40036
NC1=NC=NN1
Safety and Handling

Safety and Handling

GHS H Statement:
May cause damage to organs through prolonged or repeated exposure.
Toxic to aquatic life with long lasting effects.
Suspected of damaging the unborn child.

GHS P Statement:
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 200-521-5

RTECSNumber : XZ3850000

TSCA : TSCA

SDS Documents
Product Certifications