Please login to your online account to display your discounted pricing

3-Amino-1H-1,2,4-triazole-5-carboxylic acid hydrate, 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: fisher scientific acros  249300250

 View more versions of this product

Catalog No. AC249300250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5% (on dry substance)
Physical Form Crystalline Powder and/or Chunks
Formula Weight 128.09
Quantity 25g
Color Off-white
Melting Point 178° to 183°C
Chemical Name or Material 3-Amino-1H-1, 2, 4-triazole-5-carboxylic acid hydrate
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula C3H4N4O2·xH2O
MDL Number MFCD00149595
CAS 3641-13-2
Synonym 5-Amino-2H-1, 2, 4-triazole-3-carboxylic acid hydrate
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 3641-13-2
Loss on Drying 2% max.
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 3-Amino-1H-1, 2, 4-triazole-5-carboxylic acid hydrate
CAS 3641-13-2
CAS 3641-13-2
Assay ≥97.5% (on dry substance)
Assay Percent Range 98%
Molecular Formula C3H4N4O2·xH2O
Formula Weight 128.09
MDL Number MFCD00149595
Synonym 5-Amino-2H-1, 2, 4-triazole-3-carboxylic acid hydrate
Accessories

Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents