missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Amino-1-propanol, 99%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 104460025

Catalog No. AC104460025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

3-Amino-1-propanol
156-87-6
100.0
187.0°C to 188.0°C
Authentic
Glass bottle
1.4595 to 1.4615
MFCD00008223
04, 288
3-aminopropanol, 3-amino-1-propanol, propanolamine, 1-propanol, 3-amino, 3-propanolamine, 1-amino-3-propanol, 3-hydroxypropylamine, 3-aminopropyl alcohol, 1,3-propanolamine, beta-alaninol
WUGQZFFCHPXWKQ-UHFFFAOYSA-N
3-aminopropan-1-ol
9086
75.11
99%
98.5
0.9820g/mL
101°C
98.5% min. (GC)
C3H9NO
H2N(CH2)3OH
2.5L
0.982
Solubility in water: freely soluble. Other solubilities: miscible with alcohols,esters, chloroform, acetone, slightly soluble in arom. and aliph. hydrocarbons
C(CN)CO
75.11
36 mPa.s (20°C)
99%
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water

GHS Signal Word: Danger

EINECSNumber : 205-864-4

RTECSNumber : UA5600000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only