Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Amino-1-phenylbutane, 98%

Chemical Identifiers

Chemical Name or Material 3-Amino-1-phenylbutane
CAS 22374-89-6
Molecular Formula C10H15N
Formula Weight 149.24
MDL Number MFCD00008090
UN Number UN2922
Beilstein 2413110
Synonym 1-Methyl-3-phenylpropylamine; 4-Phenyl-2-butylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1880414

  SDS
alfa aesar™
A1880414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1880422

  SDS
alfa aesar™
A1880422
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1880436

  SDS
alfa aesar™
A1880436
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 149.24
Odor Amine-like
Melting Point −50°C
RTECS Number UI3920000
Synonym 1-Methyl-3-phenylpropylamine; 4-Phenyl-2-butylamine
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
CAS 22374-89-6
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
Chemical Name or Material 3-Amino-1-phenylbutane
MDL Number MFCD00008090
Molecular Formula C10H15N
Flash Point 97°C (206°F)
Beilstein 2413110
Quantity 25g
Boiling Point 220° to 222°C
Assay 98%
UN Number UN2922
Health Hazard 2 GHS H Statement
H301-H311-H314-H318
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.

Applications
It is employed as a chemical intermediate. 1-Methyl-3-phenylpropylamine is used in the synthesis of N-substituted derivatives of (1-methyl-3- phenylpropyl)amine.

Solubility
Slightly soluble in water. Soluble in chloroform and ethyl acetate.

Notes
Air Sensitive. Store away from air, oxidizing agents and carbon dioxide. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.

Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301-H311-H314-H318
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.