Learn More
3-Amino-1-methyl-1H-pyrazole, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H3093506
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Slightly soluble in water.
Notes
Air Sensitive, store away from air. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
3-Amino-1-methyl-1H-pyrazole | |
90°C to 92°C (1 mmHg) | |
MFCD00466340 | |
UN2735 | |
1-methyl-1h-pyrazol-3-amine, 3-amino-1-methylpyrazole, 1-methyl-3-aminopyrazole, 3-amino-1-methyl-1h-pyrazole, 1-methyl-1h-pyrazol-3-ylamine, 3-amino-1-methyl pyrazole, 1h-pyrazol-3-amine, 1-methyl, 1-methylpyrazole-3-ylamine, 3-amino-1-methyl-pyrazole, 3-amino-1 methyl pyrazole | |
MOGQNVSKBCVIPW-UHFFFAOYSA-N | |
1-methylpyrazol-3-amine | |
137254 | |
97% |
1904-31-0 | |
C4H7N3 | |
5 g | |
Air Sensitive | |
Slightly soluble in water. | |
CN1C=CC(=N1)N | |
97.121 | |
97.12 |
Chemical Identifiers
1904-31-0 | |
97.121 | |
MOGQNVSKBCVIPW-UHFFFAOYSA-N | |
137254 | |
CN1C=CC(=N1)N |
C4H7N3 | |
MFCD00466340 | |
1-methyl-1h-pyrazol-3-amine, 3-amino-1-methylpyrazole, 1-methyl-3-aminopyrazole, 3-amino-1-methyl-1h-pyrazole, 1-methyl-1h-pyrazol-3-ylamine, 3-amino-1-methyl pyrazole, 1h-pyrazol-3-amine, 1-methyl, 1-methylpyrazole-3-ylamine, 3-amino-1-methyl-pyrazole, 3-amino-1 methyl pyrazole | |
1-methylpyrazol-3-amine |
Safety and Handling
GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only