missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Amino-1-bromoisoquinoline, 97%, Thermo Scientific™

$659.08 - $659.08

Chemical Identifiers

CAS 13130-79-5
Molecular Formula C9H7BrN2
Molecular Weight (g/mol) 223.07
InChI Key PSQUIUNIVDKHJK-UHFFFAOYSA-N
Synonym 1-bromanylisoquinolin-3-amine, 1-bromo-3-aminoisoquinoline, 1-bromo-3-isoquinolinamine, 1-bromo-3-isoquinolylamine, 1-bromoisoquinolin-3-ylamine, 1-bromoisoquinoline-3-amine, 3-amino-1-bromo-isoquinoline, 3-amino-1-bromoisoquinoline, 3-isoquinolinamine, 1-bromo, acmc-1by0f
PubChem CID 289845
IUPAC Name 1-bromoisoquinolin-3-amine
SMILES C1=CC=C2C(=C1)C=C(N=C2Br)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC449160010
SDS View Documents Promotion Details
Thermo Fisher Scientific
AC449160010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC449160050
SDS View Documents Promotion Details
Thermo Fisher Scientific
AC449160050
5g
Each for $659.08
 
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

13130-79-5
223.07
1-bromanylisoquinolin-3-amine, 1-bromo-3-aminoisoquinoline, 1-bromo-3-isoquinolinamine, 1-bromo-3-isoquinolylamine, 1-bromoisoquinolin-3-ylamine, 1-bromoisoquinoline-3-amine, 3-amino-1-bromo-isoquinoline, 3-amino-1-bromoisoquinoline, 3-isoquinolinamine, 1-bromo, acmc-1by0f
1-bromoisoquinolin-3-amine
C9H7BrN2
PSQUIUNIVDKHJK-UHFFFAOYSA-N
289845
C1=CC=C2C(=C1)C=C(N=C2Br)N

Specifications

13130-79-5
96% min. (HPLC)
Glass bottle
1g
C1=CC=C2C(=C1)C=C(N=C2Br)N
223.07
223.07
3-Amino-1-bromoisoquinoline
144°C to 146°C
C9H7BrN2
1-bromanylisoquinolin-3-amine, 1-bromo-3-aminoisoquinoline, 1-bromo-3-isoquinolinamine, 1-bromo-3-isoquinolylamine, 1-bromoisoquinolin-3-ylamine, 1-bromoisoquinoline-3-amine, 3-amino-1-bromo-isoquinoline, 3-amino-1-bromoisoquinoline, 3-isoquinolinamine, 1-bromo, acmc-1by0f
PSQUIUNIVDKHJK-UHFFFAOYSA-N
1-bromoisoquinolin-3-amine
289845
97%
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash

SDS
Product Certifications

RUO – Research Use Only