Learn More
3-Amino-1-bromoisoquinoline, 97%, Thermo Scientific™
Manufacturer: Thermo Fisher Scientific AC449160050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
3-Amino-1-bromoisoquinoline | |
144.0°C to 146.0°C | |
C9H7BrN2 | |
1-bromanylisoquinolin-3-amine, 1-bromo-3-aminoisoquinoline, 1-bromo-3-isoquinolinamine, 1-bromo-3-isoquinolylamine, 1-bromoisoquinolin-3-ylamine, 1-bromoisoquinoline-3-amine, 3-amino-1-bromo-isoquinoline, 3-amino-1-bromoisoquinoline, 3-isoquinolinamine, 1-bromo, acmc-1by0f | |
PSQUIUNIVDKHJK-UHFFFAOYSA-N | |
1-bromoisoquinolin-3-amine | |
289845 | |
97% |
13130-79-5 | |
96% min. (HPLC) | |
Glass Bottle | |
5g | |
C1=CC=C2C(=C1)C=C(N=C2Br)N | |
223.07 | |
223.07 |
Chemical Identifiers
13130-79-5 | |
223.07 | |
1-bromanylisoquinolin-3-amine, 1-bromo-3-aminoisoquinoline, 1-bromo-3-isoquinolinamine, 1-bromo-3-isoquinolylamine, 1-bromoisoquinolin-3-ylamine, 1-bromoisoquinoline-3-amine, 3-amino-1-bromo-isoquinoline, 3-amino-1-bromoisoquinoline, 3-isoquinolinamine, 1-bromo, acmc-1by0f | |
1-bromoisoquinolin-3-amine |
C9H7BrN2 | |
PSQUIUNIVDKHJK-UHFFFAOYSA-N | |
289845 | |
C1=CC=C2C(=C1)C=C(N=C2Br)N |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash
RUO – Research Use Only