missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Amino-1-bromoisoquinoline, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC449160010
Specifications
3-Amino-1-bromoisoquinoline | |
144°C to 146°C | |
13130-79-5 | |
C9H7BrN2 | |
PSQUIUNIVDKHJK-UHFFFAOYSA-N | |
1-bromoisoquinolin-3-amine | |
289845 | |
97% |
Glass bottle | |
1g | |
96% min. (HPLC) | |
1-bromanylisoquinolin-3-amine, 1-bromo-3-aminoisoquinoline, 1-bromo-3-isoquinolinamine, 1-bromo-3-isoquinolylamine, 1-bromoisoquinolin-3-ylamine, 1-bromoisoquinoline-3-amine, 3-amino-1-bromo-isoquinoline, 3-amino-1-bromoisoquinoline, 3-isoquinolinamine, 1-bromo, acmc-1by0f | |
C1=CC=C2C(=C1)C=C(N=C2Br)N | |
223.07 | |
223.07 |
Chemical Identifiers
13130-79-5 | |
223.07 | |
1-bromanylisoquinolin-3-amine, 1-bromo-3-aminoisoquinoline, 1-bromo-3-isoquinolinamine, 1-bromo-3-isoquinolylamine, 1-bromoisoquinolin-3-ylamine, 1-bromoisoquinoline-3-amine, 3-amino-1-bromo-isoquinoline, 3-amino-1-bromoisoquinoline, 3-isoquinolinamine, 1-bromo, acmc-1by0f | |
1-bromoisoquinolin-3-amine |
C9H7BrN2 | |
PSQUIUNIVDKHJK-UHFFFAOYSA-N | |
289845 | |
C1=CC=C2C(=C1)C=C(N=C2Br)N |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash