Please login to your online account to display your discounted pricing

Alfa Aesar™ (+/-)-3-Amino-1-(benzyloxycarbonyl)pyrrolidine

$73.57 - $663.43

Chemical Identifiers

Chemical Name or Material (+/-)-3-Amino-1-(benzyloxycarbonyl)pyrrolidine
CAS 185057-50-5
Molecular Formula C12H16N2O2
Formula Weight 220.27
MDL Number MFCD03001673
Synonym 1-Cbz-3-aminopyrrolidine; 1-Benzyloxycarbonyl-3-aminopyrrolidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2852503

 
alfa aesar™
H2852503
1g Each for $201.65

AAH2852506

 
alfa aesar™
H2852506
5g Each for $663.43

AAH28525MD

 
alfa aesar™
H28525MD
250mg Each for $73.57
Description & Specifications

Specifications

Formula Weight 220.27
Chemical Name or Material (+/-)-3-Amino-1-(benzyloxycarbonyl)pyrrolidine
Molecular Formula C12H16N2O2
MDL Number MFCD03001673
Synonym 1-Cbz-3-aminopyrrolidine; 1-Benzyloxycarbonyl-3-aminopyrrolidine
Quantity 1g
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 185057-50-5
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.