missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Amino-1-adamantanol, ≥98%

$67.23 - $1,065.61

Chemical Identifiers

CAS 702-82-9
Molecular Formula C10H17NO
Molecular Weight (g/mol) 167.252
MDL Number MFCD01821204
InChI Key DWPIPTNBOVJYAD-UHFFFAOYSA-N
Synonym 3-amino-1-adamantanol, 3-amino-1-hydroxyadamantane, 1-amino-3-adamantanol, 3-amino-adamantanol, 1-aminoadamantane-3-ol, 3-hydroxy-1-aminoadamantane, 1-amino-3-hydroxyadamantane, tricyclo 3.3.1.13,7 decan-1-ol, 3-amino, 3-azanyladamantan-1-ol, 3-hydroxyadamantane amine
PubChem CID 658645
IUPAC Name 3-aminoadamantan-1-ol
SMILES C1C2CC3(CC1CC(C2)(C3)O)N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6413706 View Documents Alfa Aesar™
H6413706
5g Each for $67.23
 
AAH6413714 View Documents Alfa Aesar™
H6413714
25g Each for $268.32
 
AAH6413722 View Documents Alfa Aesar™
H6413722
100g Each for $1,065.61
 
Description

Description

Applications
It is employed in the synthesis of inhibitors with antihyperglycemic properties. vil- dagliptin is synthesised from 3-Amino-1-adamantanol.

Solubility
Soluble in organic solvents,insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Chemical Identifiers

702-82-9
167.252
DWPIPTNBOVJYAD-UHFFFAOYSA-N
658645
C1C2CC3(CC1CC(C2)(C3)O)N
C10H17NO
MFCD01821204
3-amino-1-adamantanol, 3-amino-1-hydroxyadamantane, 1-amino-3-adamantanol, 3-amino-adamantanol, 1-aminoadamantane-3-ol, 3-hydroxy-1-aminoadamantane, 1-amino-3-hydroxyadamantane, tricyclo 3.3.1.13,7 decan-1-ol, 3-amino, 3-azanyladamantan-1-ol, 3-hydroxyadamantane amine
3-aminoadamantan-1-ol

Specifications

265°C (literature)
702-82-9
MFCD01821204
3-amino-1-adamantanol, 3-amino-1-hydroxyadamantane, 1-amino-3-adamantanol, 3-amino-adamantanol, 1-aminoadamantane-3-ol, 3-hydroxy-1-aminoadamantane, 1-amino-3-hydroxyadamantane, tricyclo 3.3.1.13,7 decan-1-ol, 3-amino, 3-azanyladamantan-1-ol, 3-hydroxyadamantane amine
C1C2CC3(CC1CC(C2)(C3)O)N
167.252
167.25
≥98%
5g
C10H17NO
2802127
DWPIPTNBOVJYAD-UHFFFAOYSA-N
3-aminoadamantan-1-ol
658645
Solid
3-Amino-1-adamantanol
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning