Learn More
3-Amino-1-adamantanol, ≥98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6413714
Description
It is employed in the synthesis of inhibitors with antihyperglycemic properties. vil- dagliptin is synthesised from 3-Amino-1-adamantanol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is employed in the synthesis of inhibitors with antihyperglycemic properties. vil- dagliptin is synthesised from 3-Amino-1-adamantanol.
Solubility
Soluble in organic solvents,insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
3-Amino-1-adamantanol | |
265°C (literature) | |
C10H17NO | |
25 g | |
3-amino-1-adamantanol, 3-amino-1-hydroxyadamantane, 1-amino-3-adamantanol, 3-amino-adamantanol, 1-aminoadamantane-3-ol, 3-hydroxy-1-aminoadamantane, 1-amino-3-hydroxyadamantane, tricyclo 3.3.1.13,7 decan-1-ol, 3-amino, 3-azanyladamantan-1-ol, 3-hydroxyadamantane amine | |
DWPIPTNBOVJYAD-UHFFFAOYSA-N | |
3-aminoadamantan-1-ol | |
658645 | |
≥98% |
702-82-9 | |
>110°C (230°F) | |
MFCD01821204 | |
2802127 | |
Soluble in organic solvents,insoluble in water. | |
C1C2CC3(CC1CC(C2)(C3)O)N | |
167.252 | |
167.25 | |
Solid |
Chemical Identifiers
702-82-9 | |
167.252 | |
DWPIPTNBOVJYAD-UHFFFAOYSA-N | |
658645 | |
C1C2CC3(CC1CC(C2)(C3)O)N |
C10H17NO | |
MFCD01821204 | |
3-amino-1-adamantanol, 3-amino-1-hydroxyadamantane, 1-amino-3-adamantanol, 3-amino-adamantanol, 1-aminoadamantane-3-ol, 3-hydroxy-1-aminoadamantane, 1-amino-3-hydroxyadamantane, tricyclo 3.3.1.13,7 decan-1-ol, 3-amino, 3-azanyladamantan-1-ol, 3-hydroxyadamantane amine | |
3-aminoadamantan-1-ol |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only