Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Amino-1,2,4-triazole-5-carboxylic acid hydrate, 98%

$91.92 - $394.77

Chemical Identifiers

Chemical Name or Material 3-Amino-1,2,4-triazole-5-carboxylic acid hydrate
CAS 304655-78-5
Molecular Formula C3H4N4O2·xH2O
Formula Weight 128.09 (Anhydrous)
MDL Number MFCD06795815
Beilstein 124734
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1469018

  SDS
alfa aesar™
A1469018
50g Each for $91.92

AAA1469030

  SDS
alfa aesar™
A1469030
250g Each for $394.77
Description & Specifications

Specifications

Formula Weight 128.09 (Anhydrous)
Chemical Name or Material 3-Amino-1,2,4-triazole-5-carboxylic acid hydrate
TSCA TSCA
Assay 98%
Molecular Formula C3H4N4O2·xH2O
Melting Point ≈182°C (dec.)
MDL Number MFCD06795815
CAS 304655-78-5
Quantity 50g
Beilstein 124734
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
3-Amino-1,2,4-triazole-5-carboxylic acid hydrate (AmTAZAc) has been used in the synthesis of new three-dimensional metal-organic framework, [ZnF(AmTAZ)] solvents.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.