missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Amino-1,2,4-triazole-5-carboxylic acid hydrate, 98%

Manufacturer:  Alfa Aesar™ A1469018

 View more versions of this product

Catalog No. AAA1469018



Description

Description

Applications
3-Amino-1,2,4-triazole-5-carboxylic acid hydrate (AmTAZAc) has been used in the synthesis of new three-dimensional metal-organic framework, [ZnF(AmTAZ)] solvents.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications

Specifications

3-Amino-1,2,4-triazole-5-carboxylic acid hydrate
50g
C3H6N4O3
124734
AGJCUQSNRFXRDU-UHFFFAOYSA-N
3-amino-1H-1,2,4-triazole-5-carboxylic acid;hydrate
16211818
98%
~182°C (decomposition)
304655-78-5
MFCD06795815
3-amino-1,2,4-triazole-5-carboxylic acid hydrate, 1h-1,2,4-triazole-3-carboxylicacid, 5-amino-, hydrate 1:1, 3-amino-5-carboxy-1h-1,2,4-triazole hydrate, 5-amino-2h-1,2,4-triazole-3-carboxylic acid hydrate, 3-amino-1,2,4-triazole-5-carboxylic acid hemihydrate, 3-amino-1h-1,2,4-triazole-5-carboxylic acid hydrate, 3-amino-1h-1,2,4-triazole-5-carboxylic acid, oxamethane, 5-amino-1 h-1,2,4-triazole-3-carboxylic acid, 5-amino-4 h-1,2,4 triazole-3-carboxylic acid, 3-amino-1,2,4-triazole-5-carboxylic acid hydrate
C1(=NC(=NN1)N)C(=O)O.O
146.106
128.09 (Anhydrous)

Chemical Identifiers

304655-78-5
146.106
AGJCUQSNRFXRDU-UHFFFAOYSA-N
16211818
C1(=NC(=NN1)N)C(=O)O.O
C3H6N4O3
MFCD06795815
3-amino-1,2,4-triazole-5-carboxylic acid hydrate, 1h-1,2,4-triazole-3-carboxylicacid, 5-amino-, hydrate 1:1, 3-amino-5-carboxy-1h-1,2,4-triazole hydrate, 5-amino-2h-1,2,4-triazole-3-carboxylic acid hydrate, 3-amino-1,2,4-triazole-5-carboxylic acid hemihydrate, 3-amino-1h-1,2,4-triazole-5-carboxylic acid hydrate, 3-amino-1h-1,2,4-triazole-5-carboxylic acid, oxamethane, 5-amino-1 h-1,2,4-triazole-3-carboxylic acid, 5-amino-4 h-1,2,4 triazole-3-carboxylic acid, 3-amino-1,2,4-triazole-5-carboxylic acid hydrate
3-amino-1H-1,2,4-triazole-5-carboxylic acid;hydrate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents