missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Amino-1-(2,4,6-trichlorophenyl)-5-pyrazolone , 97%

$158.84 - $471.89

Chemical Identifiers

CAS 27241-31-2
Molecular Formula C9H6Cl3N3O
Molecular Weight (g/mol) 278.517
MDL Number MFCD00020743
InChI Key FOGGKKWSFIESMZ-UHFFFAOYSA-N
Synonym 3-amino-1-2,4,6-trichlorophenyl-1h-pyrazol-5 4h-one, 3-amino-1-2,4,6-trichlorophenyl-2-pyrazolin-5-one, 3-amino-1-2,4,6-trichlorophenyl-5-pyrazolone, 5-amino-2-2,4,6-trichlorophenyl-4h-pyrazol-3-one, 1-2,4,6-trichlorophenyl-3-anilinopyrazolone, 3h-pyrazol-3-one, 5-amino-2,4-dihydro-2-2,4,6-trichlorophenyl, 5-amino-2-2,4,6-trichlorophenyl-2,4-dihydro-3h-pyrazol-3-one, 1-2,4,6-trichlorophenyl 3-amino-5-pyrazolone, 3-amino-1-2,4,6-trichlorophenyl-4,5-dihydro-1h-pyrazol-5-one, 1-2,4,6-trichlorophenyl-3-amino-5-pyrazolone
PubChem CID 270746
IUPAC Name 5-amino-2-(2,4,6-trichlorophenyl)-4H-pyrazol-3-one
SMILES C1C(=NN(C1=O)C2=C(C=C(C=C2Cl)Cl)Cl)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH66022MD View Documents Alfa Aesar™
H66022MD
250mg Each for $158.84
 
AAH6602203 View Documents Alfa Aesar™
H6602203
1g Each for $471.89
 
Specifications

Chemical Identifiers

27241-31-2
278.517
FOGGKKWSFIESMZ-UHFFFAOYSA-N
270746
C1C(=NN(C1=O)C2=C(C=C(C=C2Cl)Cl)Cl)N
C9H6Cl3N3O
MFCD00020743
3-amino-1-2,4,6-trichlorophenyl-1h-pyrazol-5 4h-one, 3-amino-1-2,4,6-trichlorophenyl-2-pyrazolin-5-one, 3-amino-1-2,4,6-trichlorophenyl-5-pyrazolone, 5-amino-2-2,4,6-trichlorophenyl-4h-pyrazol-3-one, 1-2,4,6-trichlorophenyl-3-anilinopyrazolone, 3h-pyrazol-3-one, 5-amino-2,4-dihydro-2-2,4,6-trichlorophenyl, 5-amino-2-2,4,6-trichlorophenyl-2,4-dihydro-3h-pyrazol-3-one, 1-2,4,6-trichlorophenyl 3-amino-5-pyrazolone, 3-amino-1-2,4,6-trichlorophenyl-4,5-dihydro-1h-pyrazol-5-one, 1-2,4,6-trichlorophenyl-3-amino-5-pyrazolone
5-amino-2-(2,4,6-trichlorophenyl)-4H-pyrazol-3-one

Specifications

250mg
C9H6Cl3N3O
535761
FOGGKKWSFIESMZ-UHFFFAOYSA-N
5-amino-2-(2,4,6-trichlorophenyl)-4H-pyrazol-3-one
270746
Solid
3-Amino-1-(2,4,6-trichlorophenyl)-5-pyrazolone
27241-31-2
MFCD00020743
3-amino-1-2,4,6-trichlorophenyl-1h-pyrazol-5 4h-one, 3-amino-1-2,4,6-trichlorophenyl-2-pyrazolin-5-one, 3-amino-1-2,4,6-trichlorophenyl-5-pyrazolone, 5-amino-2-2,4,6-trichlorophenyl-4h-pyrazol-3-one, 1-2,4,6-trichlorophenyl-3-anilinopyrazolone, 3h-pyrazol-3-one, 5-amino-2,4-dihydro-2-2,4,6-trichlorophenyl, 5-amino-2-2,4,6-trichlorophenyl-2,4-dihydro-3h-pyrazol-3-one, 1-2,4,6-trichlorophenyl 3-amino-5-pyrazolone, 3-amino-1-2,4,6-trichlorophenyl-4,5-dihydro-1h-pyrazol-5-one, 1-2,4,6-trichlorophenyl-3-amino-5-pyrazolone
C1C(=NN(C1=O)C2=C(C=C(C=C2Cl)Cl)Cl)N
278.517
278.52
97%