missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(Amidinothio)-1-propanesulfonic acid 96%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC390181000

 View more versions of this product

Catalog No. AC390181000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥95%
198.27
White
100.0
3-(Amidinothio)-1-propanesulfonic acid
NH2C(=NH)S(CH2)3SO3H
21668-81-5
Authentic
Crystalline Powder
100g
95.0
Glass bottle
96%
C4 H10 N2 O3 S2
MFCD00085942

Chemical Identifiers

21668-81-5
100.0
C4 H10 N2 O3 S2
198.27
3-(Amidinothio)-1-propanesulfonic acid
95.0
96%
NH2C(=NH)S(CH2)3SO3H
MFCD00085942
≥95%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger