Please login to your online account to display your discounted pricing

3-(Amidinothio)-1-propanesulfonic acid 96%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-(Amidinothio)-1-propanesulfonic acid
CAS 21668-81-5
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C4 H10 N2 O3 S2
Linear Formula NH2C(=NH)S(CH2)3SO3H
Formula Weight 198.27
MDL Number MFCD00085942
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC390181000

  SDS
thermo fisher scientific
AC390181000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥95%
Physical Form Crystalline Powder
Formula Weight 198.27
Quantity 100g
Color White
CAS Min % 95.0
CAS Max % 100.0
Packaging Glass bottle
Chemical Name or Material 3-(Amidinothio)-1-propanesulfonic acid
Assay Percent Range 96%
Molecular Formula C4 H10 N2 O3 S2
Linear Formula NH2C(=NH)S(CH2)3SO3H
MDL Number MFCD00085942
CAS 21668-81-5
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.