missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(Acetylthio)propionic Acid 98.0+%, TCI America™

$118.25 - $353.22

Chemical Identifiers

CAS 41345-70-4
Molecular Formula C5H8O3S
Molecular Weight (g/mol) 148.176
MDL Number MFCD06208418
InChI Key AYQXANXXZYKTDL-UHFFFAOYSA-N
Synonym 3-(Acetylthio)propanoic Acid
PubChem CID 262719
IUPAC Name 3-acetylsulfanylpropanoic acid
SMILES CC(=O)SCCC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A31341G
View Documents
Tci America
A31341G
1g Each for $118.25
 
A31345G
View Documents
Tci America
A31345G
5g Each for $353.22
 
Specifications

Chemical Identifiers

41345-70-4
148.176
AYQXANXXZYKTDL-UHFFFAOYSA-N
262719
CC(=O)SCCC(=O)O
C5H8O3S
MFCD06208418
3-(Acetylthio)propanoic Acid
3-acetylsulfanylpropanoic acid

Specifications

128°C
54°C
41345-70-4
MFCD06208418
AYQXANXXZYKTDL-UHFFFAOYSA-N
3-acetylsulfanylpropanoic acid
262719
Crystal-Powder at 20°C
3-(Acetylthio)propionic Acid
White-Yellow
1g
C5H8O3S
3-(Acetylthio)propanoic Acid
CC(=O)SCCC(=O)O
148.176
148.18
≥98.0% (GC,T)
Safety and Handling

Safety and Handling

TSCA : No