Please login to your online account to display your discounted pricing

3-Acetylpyridine, 98%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: fisher scientific acros  102690100

 View more versions of this product

Catalog No. AC102690100

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid
Formula Weight 121.14
Quantity 10g
Color Clear colorless to yellow
CAS Max % 100.0
Packaging Glass bottle
Infrared Spectrum Authentic
CAS 350-03-8
Health Hazard 2 GHS P Statement
Causes skin irritation.
Fatal if swallowed.
Causes serious eye irritation.
CAS Min % 97.5
Molecular Formula C7H7NO
Name Note 98%
MDL Number MFCD00006396
Assay Percent Range 98%
Linear Formula (C5H4N)COCH3
Chemical Name or Material 3-Acetylpyridine
Refractive Index 1.5320 to 1.5350 (20°C, 589nm)
Synonym Methyl 3-pyridyl ketone
Health Hazard 3 GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 GHS Signal Word: Danger

Specifications

Chemical Name or Material 3-Acetylpyridine
Name Note 98%
CAS 350-03-8
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H7NO
Linear Formula (C5H4N)COCH3
Formula Weight 121.14
MDL Number MFCD00006396
Synonym Methyl 3-pyridyl ketone
Accessories

GHS Signal Word: Danger

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Causes skin irritation.
Fatal if swallowed.
Causes serious eye irritation.

SDS & Safety Documents