missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Acetylphenylboronic acid, 96%, ACROS Organics™

Chemical Identifiers

CAS 204841-19-0
Molecular Formula C8H9BO3
Molecular Weight (g/mol) 163.97
MDL Number MFCD01074678
InChI Key SJGGDZCTGBKBCK-UHFFFAOYSA-N
Synonym 1-3-dihydroxyboranyl phenyl ethan-1-one
PubChem CID 2734310
ChEBI CHEBI:86552
IUPAC Name (3-acetylphenyl)boronic acid
SMILES B(C1=CC(=CC=C1)C(=O)C)(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC358830010 SDS View Documents Fisher Scientific Acros
358830010
1g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC358830100 SDS View Documents Thermo Fisher Scientific
358830100
10g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

204841-19-0
163.97
SJGGDZCTGBKBCK-UHFFFAOYSA-N
2734310
(3-acetylphenyl)boronic acid
C8H9BO3
MFCD01074678
1-3-dihydroxyboranyl phenyl ethan-1-one
CHEBI:86552
B(C1=CC(=CC=C1)C(=O)C)(O)O

Specifications

Authentic
Yellow
1g
95.0
95% min. (HPLC)
MFCD01074678
SJGGDZCTGBKBCK-UHFFFAOYSA-N
(3-acetylphenyl)boronic acid
2734310
163.97
96%
Glass bottle
212°C to 214°C
204841-19-0
100.0
C8H9BO3
1-3-dihydroxyboranyl phenyl ethan-1-one
B(C1=CC(=CC=C1)C(=O)C)(O)O
163.97
CHEBI:86552
Crystalline Powder
3-Acetylphenylboronic acid
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning