Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Acetylphenyl isothiocyanate, 97%

Chemical Identifiers

Chemical Name or Material 3-Acetylphenyl isothiocyanate
CAS 3125-71-1
Molecular Formula C9H7NOS
Formula Weight 177.23
MDL Number MFCD00041081
UN Number UN2923
Beilstein 2803021
Synonym 3'-Isothiocyanatoacetophenone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1013403

  SDS
alfa aesar™
L1013403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1013406

  SDS
alfa aesar™
L1013406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 177.23
CAS 3125-71-1
Chemical Name or Material 3-Acetylphenyl isothiocyanate
Molecular Formula C9H7NOS
Synonym 3'-Isothiocyanatoacetophenone
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Assay 97%
Health Hazard 1 Danger
Flash Point >110°C (230°F)
Beilstein 2803021
MDL Number MFCD00041081
Health Hazard 2 GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Melting Point ≈24°C
Quantity 1g
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
UN Number UN2923
Odor Irritating
Boiling Point 135° to 136°C (2mmHg)
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.