missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Acetylphenyl isothiocyanate, 97%

Manufacturer:  Alfa Aesar™ L1013406

 View more versions of this product

Catalog No. AAL1013406



Specifications

Specifications

3-Acetylphenyl isothiocyanate
Irritating
~24°C
3125-71-1
MFCD00041081
2803021
RGANVWCPAAIVNN-UHFFFAOYSA-N
1-(3-isothiocyanatophenyl)ethanone
520515
97%
>110°C (230°F)
135°C to 136°C (2mmHg)
5g
C9H7NOS
UN2923
3-acetylphenyl isothiocyanate, 1-3-isothiocyanatophenyl ethanone, 3-acetylphenylisothiocyanate, 1-3-isothiocyanato-phenyl-ethanone, 3-acetylbenzenisothiocyanate, acmc-20anak, 3'-isothiocyanatoacetophenone, 3-methylcarbonylphenyl isothiocyanate, 1-3-isothiocyanatophenyl ethanone #, ethanone,1-3-isothiocyanatophenyl
CC(=O)C1=CC(=CC=C1)N=C=S
177.221
177.23

Chemical Identifiers

3125-71-1
177.221
RGANVWCPAAIVNN-UHFFFAOYSA-N
520515
CC(=O)C1=CC(=CC=C1)N=C=S
C9H7NOS
MFCD00041081
3-acetylphenyl isothiocyanate, 1-3-isothiocyanatophenyl ethanone, 3-acetylphenylisothiocyanate, 1-3-isothiocyanato-phenyl-ethanone, 3-acetylbenzenisothiocyanate, acmc-20anak, 3'-isothiocyanatoacetophenone, 3-methylcarbonylphenyl isothiocyanate, 1-3-isothiocyanatophenyl ethanone #, ethanone,1-3-isothiocyanatophenyl
1-(3-isothiocyanatophenyl)ethanone
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.

Documents