Please login to your online account to display your discounted pricing

3-Acetylphenyl isocyanate, 97%, ACROS Organics™

$123.03 - $123.03

Chemical Identifiers

Chemical Name or Material 3-Acetylphenyl isocyanate
Name Note 97%
CAS 23138-64-9
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C9H7NO2
Linear Formula CH3COC6H4NCO
Formula Weight 161.16
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC314100010

  SDS
thermo fisher scientific
AC314100010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC314100100

  SDS
thermo fisher scientific
314100100
Glass bottle 10g Each for $123.03
Description & Specifications

Specifications

Physical Form Liquid After Melting
Assay ≥96.0%
Formula Weight 161.16
Quantity 1g
Color Clear yellow to light brown
TSCA TSCA
Density 1.17g/mL
CAS Min % 96.0
CAS Max % 100.0
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 97%
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Chemical Name or Material 3-Acetylphenyl isocyanate
Molecular Formula C9H7NO2
Name Note 97%
CAS 23138-64-9
Linear Formula CH3COC6H4NCO
MDL Number MFCD00013866
Infrared Spectrum Authentic
Refractive Index 1.5610 to 1.5630 (20°C, 589nm)
Packaging Glass bottle
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.