Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Acetylphenyl isocyanate, 97%

Chemical Identifiers

Chemical Name or Material 3-Acetylphenyl isocyanate
CAS 23138-64-9
Molecular Formula C9H7NO2
Formula Weight 161.16g/mol
MDL Number MFCD00013866
UN Number UN2206
Beilstein 387294
Synonym 3'-Isocyanatoacetophenone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1203003

 
alfa aesar™
L12030-03
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Boiling Point 154° to 155°C (4mmHg)
Quantity 1g
Flash Point >110°C (230°F)
Odor Irritating
Chemical Name or Material 3-Acetylphenyl isocyanate
CAS 23138-64-9
Assay 97%
Molecular Formula C9H7NO2
Formula Weight 161.16g/mol
MDL Number MFCD00013866
UN Number UN2206
Beilstein 387294
Synonym 3'-Isocyanatoacetophenone
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.