Please login to your online account to display your discounted pricing

3-Acetylphenanthrene, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Acetylphenanthrene
CAS 2039-76-1
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C16H12O
Formula Weight 220.27
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC102640050

  SDS
thermo fisher scientific
AC102640050
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC102640250

  SDS
thermo fisher scientific
AC102640250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Health Hazard 3 GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Health Hazard 2 GHS H Statement
Harmful if swallowed. Harmful in contact with skin. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Physical Form Powder
Formula Weight 220.27
Assay ≥96.0%
Quantity 5g
Color White to pale yellow
Melting Point 71° to 76°C
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Molecular Formula C16H12O
CAS 2039-76-1
Chemical Name or Material 3-Acetylphenanthrene
Assay Percent Range 97%
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
Harmful if swallowed. Harmful in contact with skin. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.