Please login to your online account to display your discounted pricing

3-Acetylphenanthrene, 97%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC102640250

 View more versions of this product

Catalog No. AC102640250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder
Assay ≥96.0%
Formula Weight 220.27
Quantity 25g
Color White to pale yellow
Melting Point 71° to 76°C
CAS Min % 96.0
CAS Max % 100.0
Packaging Glass bottle
Assay Percent Range 97%
CAS 2039-76-1
Chemical Name or Material 3-Acetylphenanthrene
Molecular Formula C16H12O
Infrared Spectrum Authentic
Health Hazard 1 Warning
Health Hazard 2 GHS P Statement Harmful if swallowed. Harmful in contact with skin. Causes skin irritation. Causes serious eye irritation.
Health Hazard 3 GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protec

Specifications

Chemical Name or Material 3-Acetylphenanthrene
CAS 2039-76-1
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C16H12O
Formula Weight 220.27
Accessories

Warning

GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protec

GHS P Statement Harmful if swallowed. Harmful in contact with skin. Causes skin irritation. Causes serious eye irritation.