Please login to your online account to display your discounted pricing

3-Acetylbenzoic acid, 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: fisher scientific acros  AC366500010

 View more versions of this product

Catalog No. AC366500010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 164.16
Quantity 1g
Color White to light brown
Melting Point 163° to 171°C
CAS Max % 100.0
Infrared Spectrum Authentic
Molecular Formula C9H8O3
Linear Formula CH3COC6H4COOH
MDL Number MFCD00045847
Packaging Glass bottle
Assay Percent Range 98%
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Chemical Name or Material 3-Acetylbenzoic acid
CAS 586-42-5
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 3-Acetylbenzoic acid
CAS 586-42-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H8O3
Linear Formula CH3COC6H4COOH
Formula Weight 164.16
MDL Number MFCD00045847
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents