Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Acetylbenzeneboronic acid, 97%

$289.38 - $289.38

Chemical Identifiers

Chemical Name or Material 3-Acetylbenzeneboronic acid
CAS 204841-19-0
Molecular Formula C8H9BO3
Formula Weight 163.97
MDL Number MFCD01074678
Beilstein 8255673
Synonym 3-Acetylphenylboronic acid; 3'-Boronoacetophenone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2347814

  SDS
alfa aesar™
B2347814
25g Each for $289.38
Description & Specifications

Specifications

Formula Weight 163.97
Chemical Name or Material 3-Acetylbenzeneboronic acid
Assay 97%
Molecular Formula C8H9BO3
MDL Number MFCD01074678
CAS 204841-19-0
Beilstein 8255673
Synonym 3-Acetylphenylboronic acid; 3'-Boronoacetophenone
Health Hazard 1 Warning
Melting Point ≈200° to 205°C
Quantity 25g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.