Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Acetylacrylic acid, 97%

$134.58 - $134.58

Chemical Identifiers

Chemical Name or Material 3-Acetylacrylic acid
CAS 4743-82-2
Molecular Formula C5H6O3
Formula Weight 114.10g/mol
MDL Number MFCD00014016
Beilstein 1745249
Synonym 4-Ketopent-2-enoic acid; 4-Oxopent-2-enoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0218503

 
alfa aesar™
L02185-03
1g Each for $134.58
Description & Specifications

Specifications

Melting Point 122° to 126°C
Quantity 1g
Chemical Name or Material 3-Acetylacrylic acid
CAS 4743-82-2
Assay 97%
Molecular Formula C5H6O3
Formula Weight 114.10g/mol
MDL Number MFCD00014016
Beilstein 1745249
Synonym 4-Ketopent-2-enoic acid; 4-Oxopent-2-enoic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
RTECS Number SB3360000
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C