Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Acetyl-2-methyl-5-phenylthiophene, 98%

$227.81 - $227.81

Chemical Identifiers

Chemical Name or Material 3-Acetyl-2-methyl-5-phenylthiophene
CAS 40932-63-6
Molecular Formula C13H12OS
Formula Weight 216.3
MDL Number MFCD00151790
Beilstein 8481276
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1355406

  SDS
alfa aesar™
L1355406
5g Each for $227.81
Description & Specifications

Specifications

Formula Weight 216.3
Assay 98%
Molecular Formula C13H12OS
MDL Number MFCD00151790
CAS 40932-63-6
Chemical Name or Material 3-Acetyl-2-methyl-5-phenylthiophene
Beilstein 8481276
Melting Point 67° to 69°C
Quantity 5g