Alfa Aesar™ 3-Acetyl-2-methyl-5-phenylthiophene, 98%
Manufacturer:  Alfa Aesar™ L1355403
 View more versions of this product
Catalog No. AAL1355403
Specifications
Specifications
3-Acetyl-2-methyl-5-phenylthiophene
1g
C13H12OS
8481276
GREAZYFTAJMZFD-UHFFFAOYSA-N
1-(2-methyl-5-phenylthiophen-3-yl)ethanone
2728765
98%
67°C to 69°C
40932-63-6
MFCD00151790
3-acetyl-2-methyl-5-phenylthiophene, 1-2-methyl-5-phenylthiophen-3-yl ethanone, 1-2-methyl-5-phenylthien-3-yl ethanone, ethanone, 1-2-methyl-5-phenyl-3-thienyl, acmc-20anao, 3-acetyl-2-methyl-5-phenylthiohene, 1-2-methyl-5-phenyl-3-thienyl ethanone, 1-2-methyl-5-phenyl-3-thiophenyl ethanone, 1-2-methyl-5-phenyl-thiophen-3-yl ethanone, 1-2-methyl-5-phenyl-thiophen-3-yl-ethanone
CC1=C(C=C(S1)C2=CC=CC=C2)C(=O)C
216.298
216.3
Chemical Identifiers
40932-63-6
216.298
GREAZYFTAJMZFD-UHFFFAOYSA-N
2728765
CC1=C(C=C(S1)C2=CC=CC=C2)C(=O)C
C13H12OS
MFCD00151790
3-acetyl-2-methyl-5-phenylthiophene, 1-2-methyl-5-phenylthiophen-3-yl ethanone, 1-2-methyl-5-phenylthien-3-yl ethanone, ethanone, 1-2-methyl-5-phenyl-3-thienyl, acmc-20anao, 3-acetyl-2-methyl-5-phenylthiohene, 1-2-methyl-5-phenyl-3-thienyl ethanone, 1-2-methyl-5-phenyl-3-thiophenyl ethanone, 1-2-methyl-5-phenyl-thiophen-3-yl ethanone, 1-2-methyl-5-phenyl-thiophen-3-yl-ethanone
1-(2-methyl-5-phenylthiophen-3-yl)ethanone
Documents