Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Acetyl-2,5-dimethylfuran, 98%

$44.45 - $712.70

Chemical Identifiers

Chemical Name or Material 3-Acetyl-2,5-dimethylfuran
CAS 10599-70-9
Molecular Formula C8H10O2
Formula Weight 138.17
MDL Number MFCD00010281
Beilstein 112094
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1554509

  SDS
alfa aesar™
A1554509
10g Each for $44.45

AAA1554518

  SDS
alfa aesar™
A1554518
50g Each for $174.78

AAA1554530

  SDS
alfa aesar™
A1554530
250g Each for $712.70
Description & Specifications

Specifications

Formula Weight 138.17
Assay 98%
Health Hazard 1 Danger
Molecular Formula C8H10O2
Quantity 10g
Flash Point 78°C (172°F)
Beilstein 112094
Health Hazard 2 GHS H Statement
H301-H227-H319-H317
Toxic if swallowed.
Combustible liquid.
Causes serious eye irritation.
May cause an allergic skin reaction.
CAS 10599-70-9
Health Hazard 3 GHS P Statement
P210-P261-P301+P310-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 3-Acetyl-2,5-dimethylfuran
MDL Number MFCD00010281
Boiling Point 198°C
Safety and Handling

Danger

GHS P Statement
P210-P261-P301+P310-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H227-H319-H317
Toxic if swallowed.
Combustible liquid.
Causes serious eye irritation.
May cause an allergic skin reaction.